missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) fluoride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
50 g
Description
It is used as a catalyst to alloy metals and also used for optical deposition, of which it tremendously improves optical quality, in dental care.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1400°C |
Melting Point | 1200°C |
Physical Form | Powder |
Quantity | 2 g |
UN Number | UN2923 |
Merck Index | 14,2440 |
Solubility Information | Soluble in water,alcohol and acetone. |
Formula Weight | 96.93 |
Percent Purity | 99.99% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'