missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) gluconate hydrate, Thermo Scientific Chemicals

Product Code. 11485707
Click to view available options
Quantity:
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1809162-99-9
Molecular Formula C12H26CoO15
Molecular Weight (g/mol) 469.258
MDL Number MFCD02684006
InChI Key QBVYGKYWWCNZSO-XRDLMGPZSA-N
Synonym cobalt ii gluconate hydrate, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoyl oxy cobaltio 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate
PubChem CID 117064710
IUPAC Name cobalt;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;hydrate
SMILES C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.[Co]

Specifications

Melting Point >300°C
Quantity 100 g
Formula Weight 449.23 (Anhydrous)
Chemical Name or Material Cobalt(II) gluconate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'