missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hydroxide, 99.9% (metals basis)

Product Code. 11361859
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11361859

Brand: Thermo Scientific Alfa Aesar 012524.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21041-93-0 | CoH2O2 | 92.95 g/mol

Cobalt(II) hydroxide is used as a drying agent for paints and varnishes. It acts as an aid to dry the lithographic inks and catalyst in the preparation of battery electrode impregnating solutions. It is also involved in the synthesis of other cobalt compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21041-93-0
Molecular Formula CoH2O2
Molecular Weight (g/mol) 92.95
MDL Number MFCD00016015
InChI Key ASKVAEGIVYSGNY-UHFFFAOYSA-L
Synonym cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
PubChem CID 10129900
IUPAC Name cobalt(2+);dihydroxide
SMILES [OH-].[OH-].[Co++]

Specifications

Melting Point 250°C (decomposition)
Color Pink to Purple
Physical Form Powder
Quantity 500 g
Linear Formula Co(OH)2
Merck Index 14,2442
Solubility Information Soluble in acids and ammonia; Very slightly soluble in water; Insoluble in dilute alkalis.
Formula Weight 92.95
Percent Purity 99.9%
Odor Odorless
Sensitivity Air Sensitive
Assay (metals basis)
Chemical Name or Material Cobalt(II) hydroxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.