Learn More
Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Description
Finds use in enamels, semiconductors, grinding wheels. Used in catalysis, superconductors, ceramics and other fields as an important inorganic material. Also used as catalyst and catalyst carrier, as electrode active material, for glass, porcelain colorants and pigments, as chemical industry oxidant and as a catalyst for organic synthesis, in senior goggles and other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices, enamels, solar energy absorbers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 900°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.7% (Metals basis) |
Solubility Information | Soluble in acids and alkalis. Insoluble in water. |
IUPAC Name | oxocobalt;oxo(oxocobaltiooxy)cobalt |
Formula Weight | 240.8 |
Odor | Odorless |
Packaging | Packing group III |
Mehr anzeigen |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q03J024 | Certificate of Analysis | 09/05/2023 | 040184.22 |
Q18J019 | Certificate of Analysis | 25/04/2023 | 040184.22 |
Z21I032 | Certificate of Analysis | 05/01/2023 | 040184.14,040184.36,040184.A4,040184.22 |
T26I015 | Certificate of Analysis | 02/08/2022 | 040184.14,040184.36,040184.A4,040184.22 |
S27I018 | Certificate of Analysis | 11/07/2022 | 040184.14,040184.36,040184.A4,040184.22 |
Sicherheit und Handhabung



- Cobalt(II,III) oxide
- Warning
- Carcinogenicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.