missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, 97.7% min

Product Code. 11351109
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
25g
5kg
This item is not returnable. View return policy

Product Code. 11351109

Brand: Thermo Scientific Alfa Aesar 011341.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol

Cobalt(II) nitrate hexahydrate is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C to 56°C
Color Red
Physical Form Crystalline
Quantity 1 kg
UN Number UN1477
Merck Index 14,2444
Solubility Information Soluble in water,alcohol,acetone and ammonia.
Formula Weight 291.04 (182.94 Anhydrous)
Percent Purity ≤97.7%
Odor Odorless
Refractive Index 1.52
Density 1.87 g/mL
Chemical Name or Material Cobalt(II) nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Oxidising solid Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Cobalt(II) nitrate,hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.