missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, 99%, pure, Thermo Scientific Chemicals

Product Code. 10388010
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55.0°C
Color Brown-Red to Pink
Physical Form Adhering Crystalline Powder
Quantity 100 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Co(NO3)2·6H2O
Merck Index 15, 2428
Solubility Information Solubility in water: 1340g/L. Other solubilities: soluble in ethanol and acetone, soluble in most common organic solvents
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
Formula Weight 291.02
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Specific Gravity 1.88
Density 1.8800g/mL
Chemical Name or Material Cobalt(II) nitrate hexahydrate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.