missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(Ii) Nitrate Hexahydrate, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 5 g |
Linear Formula | Co(NO3)2 · 6H2O |
UN Number | 1477 |
Formula Weight | 291.03g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Chemical Name or Material | Cobalt(Ii) Nitrate Hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'