missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxalate, anhydrous, 98%, Thermo Scientific Chemicals

£31.10 - £103.00

Chemical Identifiers

CAS 814-89-1
Molecular Formula C2CoO4
Molecular Weight (g/mol) 146.951
MDL Number MFCD00050793
InChI Key MULYSYXKGICWJF-UHFFFAOYSA-L
Synonym cobalt oxalate, cobalt oxalate 1:1, cobalt oxalate coc2o4, cobalt; oxalic acid, oxalic acid cobalt 2+ salt 1:1, ethanedioic acid, cobalt 2+ salt 1:1, oxalic acid cobalt, cobalt ii oxalate,anhydrous, oxalic acid, cobalt 2+ salt 1:1
PubChem CID 69946
IUPAC Name cobalt(2+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Co+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11456106
View Documents
Thermo Scientific Alfa Aesar
B22956.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11466106
View Documents
Thermo Scientific Alfa Aesar
B22956.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

814-89-1
146.951
MULYSYXKGICWJF-UHFFFAOYSA-L
69946
C(=O)(C(=O)[O-])[O-].[Co+2]
C2CoO4
MFCD00050793
cobalt oxalate, cobalt oxalate 1:1, cobalt oxalate coc2o4, cobalt; oxalic acid, oxalic acid cobalt 2+ salt 1:1, ethanedioic acid, cobalt 2+ salt 1:1, oxalic acid cobalt, cobalt ii oxalate,anhydrous, oxalic acid, cobalt 2+ salt 1:1
cobalt(2+);oxalate

Specifications

>300°C
C2CoO4
14,2445
MULYSYXKGICWJF-UHFFFAOYSA-L
cobalt(2+);oxalate
69946
98%
Cobalt(II) oxalate, anhydrous
814-89-1
MFCD00050793
cobalt oxalate, cobalt oxalate 1:1, cobalt oxalate coc2o4, cobalt; oxalic acid, oxalic acid cobalt 2+ salt 1:1, ethanedioic acid, cobalt 2+ salt 1:1, oxalic acid cobalt, cobalt ii oxalate,anhydrous, oxalic acid, cobalt 2+ salt 1:1
C(=O)(C(=O)[O-])[O-].[Co+2]
146.951
146.95
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c

H302+H312-H350

missing translation for 'einecsNumber' : 212-409-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only