Learn More
Cobalt(II) oxide, 95%, Thermo Scientific Chemicals
£77.10 - £234.00
Chemical Identifiers
CAS | 1307-96-6 |
---|---|
Molecular Formula | CoO |
Molecular Weight (g/mol) | 74.93 |
MDL Number | MFCD00016031 |
InChI Key | IVMYJDGYRUAWML-UHFFFAOYSA-N |
Synonym | cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 |
PubChem CID | 14786 |
IUPAC Name | oxocobalt |
SMILES | O=[Co] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392608
|
Thermo Scientific Alfa Aesar
023141.14 |
25 g |
£77.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302618
|
Thermo Scientific Alfa Aesar
023141.22 |
100 g |
£234.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example, in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
1307-96-6 | |
74.93 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
14786 | |
O=[Co] |
CoO | |
MFCD00016031 | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
oxocobalt |
Specifications
1935°C | |
Powder | |
CoO | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
oxocobalt | |
14786 | |
Air Sensitive | |
Cobalt(II) oxide |
1307-96-6 | |
95% | |
MFCD00016031 | |
Soluble in acids or alkalis. Insoluble in water. | |
O=[Co] | |
74.93 | |
74.93 | |
6.45 |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P301+P310-P302+P352-P304+P340-P308+P313-P310-P330-P333+P313-P363-P501c
H301-H317-H330-H334-H350
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 215-154-6
missing translation for 'rtecsNumber' : GG2800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only