Learn More
Cobalt(II) oxide, 99.995% (metals basis), Thermo Scientific Chemicals
£123.00 - £1449.00
Chemical Identifiers
CAS | 1307-96-6 |
---|---|
Molecular Formula | CoO |
Molecular Weight (g/mol) | 74.93 |
MDL Number | MFCD00016031 |
InChI Key | IVMYJDGYRUAWML-UHFFFAOYSA-N |
Synonym | cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 |
PubChem CID | 14786 |
IUPAC Name | oxocobalt |
SMILES | O=[Co] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488500
|
Thermo Scientific Alfa Aesar
044354.06 |
5 g |
£123.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478500
|
Thermo Scientific Alfa Aesar
044354.14 |
25 g |
£452.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468500
|
Thermo Scientific Alfa Aesar
044354.22 |
100 g |
£1449.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) oxide is widely used in the ceramic industry. It is an additive to create blue-colored glazes and enamels. It finds application in the manufacture of magnetic and electronic component materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1307-96-6 | |
74.93 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
14786 | |
O=[Co] |
CoO | |
MFCD00016031 | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
oxocobalt |
Specifications
1935°C | |
Brown-Green | |
CoO | |
UN2811 | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
oxocobalt | |
14786 | |
99.995% | |
Air Sensitive | |
(metals basis) |
1307-96-6 | |
Powder | |
MFCD00016031 | |
14,2446 | |
Insoluble in water. | |
O=[Co] | |
74.93 | |
74.93 | |
CoO, may contain up to 10% Co3O4 | |
6.45 g/mL | |
Cobalt (II) oxide |
Safety and Handling
GHS H Statement
H351-H302-H317
Suspected of causing cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P301+P310-P302+P352-P304+P340-P308+P313-P310-P330-P333+P313-P363-P501c
H301-H317-H330-H334-H350
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 215-154-6
missing translation for 'rtecsNumber' : GG2800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only