Learn More
Cobalt(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Description
Cobalt(II) perchlorate hexahydrate is used as chemical organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Crystalline |
Quantity | 10 g |
UN Number | UN1481 |
Solubility Information | Soluble in water. |
Formula Weight | 366.74 (257.83 Anhydrous) |
Grade | Reagent |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Cobalt(II) perchlorate hexahydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X15K047 | Certificate of Analysis | 30/10/2024 | 011652.09,011652.18,011652.30 |
Q28I036 | Certificate of Analysis | 17/11/2022 | 011652.09,011652.18,011652.30 |
K20S065 | Certificate of Analysis | 08/12/2021 | 011652.09,011652.18,011652.30 |
W07H055 | Certificate of Analysis | 08/12/2021 | 011652.09,011652.18,011652.30 |
Q25A055 | Certificate of Analysis | 08/12/2021 | 011652.09,011652.18,011652.30 |
Safety and Handling


- cobalt perchlorate, hexahydrate reagent
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Cobalt(II) perchlorate hexahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.