Learn More
Cobalt(II) sulfamate hydrate, Thermo Scientific Chemicals
£92.76 - £891.44
Chemical Identifiers
CAS | 14017-41-5 |
---|---|
Molecular Formula | CoH4N2O6S2 |
Molecular Weight (g/mol) | 251.09 |
MDL Number | MFCD00049877 |
InChI Key | WLQXLCXXAPYDIU-UHFFFAOYSA-L |
Synonym | cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek |
PubChem CID | 26398 |
SMILES | [Co++].NS([O-])(=O)=O.NS([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384948
|
Thermo Scientific Alfa Aesar
043906.22 |
100 g |
£92.76
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304958
|
Thermo Scientific Alfa Aesar
043906.36 |
500 g |
£297.48
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394948
|
Thermo Scientific Alfa Aesar
043906.A3 |
2 kg |
£891.44
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) sulfamate hydrate is used in photonic device fabrication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14017-41-5 | |
251.09 | |
WLQXLCXXAPYDIU-UHFFFAOYSA-L | |
26398 |
CoH4N2O6S2 | |
MFCD00049877 | |
cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek | |
[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O |
Specifications
14017-41-5 | |
CoH4N2O6S2 | |
cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek | |
WLQXLCXXAPYDIU-UHFFFAOYSA-L | |
λ²-cobalt(2+) disulfamate | |
26398 | |
Odorless |
Powder | |
MFCD00049877 | |
Soluble in water. | |
[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O | |
251.09 | |
251.11 (Anhydrous) | |
Cobalt (II) sulfamate hydrate |
Safety and Handling
GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H341-H350i-H360F
missing translation for 'einecsNumber' : 237-834-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only