Learn More
Cobalt(II) sulfate heptahydrate, 99+%, extra pure, Thermo Scientific Chemicals
£39.30 - £326.63
Chemical Identifiers
CAS | 10026-24-1 |
---|---|
Molecular Formula | CoH14O11S |
Molecular Weight (g/mol) | 281.09 |
MDL Number | MFCD00149658 |
InChI Key | MEYVLGVRTYSQHI-UHFFFAOYSA-L |
Synonym | cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o |
PubChem CID | 61444 |
SMILES | O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10686812
|
Thermo Scientific Acros
213101000 |
100 g | Glass bottle |
£39.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378060
|
Thermo Scientific Acros
213105000 |
500 g | Glass bottle |
£116.58
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10711234
|
Thermo Scientific Acros
213100025 |
2.5 kg | Glass bottle |
£326.63
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
10026-24-1 | |
281.09 | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
61444 |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Specifications
98.0°C | |
Red to Brown | |
99% min. (Complexometry) | |
CoSO4·7H2O | |
15,2432 | |
Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O | |
281.09 | |
281.09 | |
Extra Pure | |
Cobalt(II) sulfate heptahydrate, 99+% |
10026-24-1 | |
Crystalline Powder and/or Crystals | |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
λ²-cobalt(2+) heptahydrate sulfate | |
61444 | |
≥99% | |
2.03 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : GG3200000
- Cobalt(II) sulfate heptahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : Cobalt(II) sulfate heptahydrate, Cobaltsulfate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.