Learn More
Cobalt(II) sulfate heptahydrate, for analysis, Thermo Scientific Chemicals
£58.90 - £190.00
Chemical Identifiers
CAS | 10026-24-1 |
---|---|
Molecular Formula | CoH14O11S |
Molecular Weight (g/mol) | 281.09 |
MDL Number | MFCD00149658 |
InChI Key | MEYVLGVRTYSQHI-UHFFFAOYSA-L |
Synonym | cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o |
PubChem CID | 61444 |
SMILES | O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15234318
|
Thermo Scientific Acros
453902500 |
250 g |
£58.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15224318
|
Thermo Scientific Acros
453900010 |
1 kg |
£190.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10026-24-1 | |
281.09 | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
61444 |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Specifications
98.0°C | |
4 to 6 (5% aq. soln.) | |
CoSO4·7H2O | |
15,2432 | |
Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O | |
281.09 | |
281.09 | |
Analytical |
10026-24-1 | |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
cobalt(2+);sulfate;heptahydrate | |
61444 | |
99.0 to 101.0% (Complexometry) | |
Cobalt(II) sulfate heptahydrate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT
GHS Signal Word: Danger
RUO – Research Use Only