missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) tetrafluoroborate hydrate, 96%, Thermo Scientific Chemicals

Product Code. 11312618
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a cobalt salt. It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26490-63-1
MDL Number MFCD00016012

Specifications

Physical Form Crystalline
Quantity 100 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 232.54 (Anhydrous)
Percent Purity 96%
Sensitivity Hygroscopic
Chemical Name or Material Cobalt(II) tetrafluoroborate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'