missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) thiocyanate, 98+%, Thermo Scientific Chemicals

Product Code. 11416420
Click to view available options
Quantity:
10 g
50 g
250 g

Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

Solubility
Soluble in water.

Notes
Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3017-60-5
Molecular Formula C2H2CoN2S2
Molecular Weight (g/mol) 177.105
MDL Number MFCD00010937
InChI Key IQHZUTZWNWRODJ-UHFFFAOYSA-N
PubChem CID 131877393
IUPAC Name cobalt;thiocyanic acid
SMILES C(#N)S.C(#N)S.[Co]

Specifications

Odor Odorless
Linear Formula Co(SCN)2
Quantity 50 g
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 175.1
Percent Purity ≥98%
Chemical Name or Material Cobalt(II) thiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'