missing translation for 'onlineSavingsMsg'
Learn More

Cobalt organometallic oil standard 5000ppm in oil, SPEX Certiprep™

Product Code. 11905184
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Quantity 50 g
compliance-icons compliance-icons
Product Identifier
  • 5000 ug/g Cobalt in Base Oil 75
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : cobalt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'