missing translation for 'onlineSavingsMsg'
Learn More
Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%, Thermo Scientific Chemicals Product Code.: 11367818

Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%, Thermo Scientific Chemicals

Product Code. 11367818
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11367818

Brand: Thermo Scientific Alfa Aesar 036375.18

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Color Gray
Quantity 50 g
Percent Purity 99.5%
Odor Odorless
Particle Size 12mm (0.47 in.) & down
Form Pieces
Assay (metals basis excluding Ni), Ni nominal 1%
Chemical Name or Material Cobalt pieces

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG28N26 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number036378.22,036375.30,036375.18
Lot NumberE14L28 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number036378.22,036375.30,036375.18
Lot NumberE28Z002 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number036378.22,036375.30,036375.18
Lot NumberG31L41 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number036378.22,036375.30,036375.18
Lot NumberC18T049 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number036378.22,036375.30,036375.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.