missing translation for 'onlineSavingsMsg'
Learn More
Cobalt powder, 1.6 micron, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11375958

Cobalt powder, 1.6 micron, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11375958
100 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11375958

Brand: Thermo Scientific Alfa Aesar 010455.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Color Black
UN Number UN3089
Quantity 100 g
Percent Purity 98.8%
Odor Odorless
Particle Size 1.6 Micron
Form Powder
Assay (metals basis)
Chemical Name or Material Cobalt powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ05K036 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number010455.22,010455.36,010455.09
Lot NumberZ23F044 Certificate TypeCertificate of Analysis Date25/04/2023 Catalog Number010455.22
Lot NumberT19I013 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog Number010455.22,010455.36,010455.09
Lot NumberN07I069 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010455.22,010455.36,010455.09
Lot NumberY16H003 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010455.22,010455.36,010455.09
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt powder
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : Cobalt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt powder, 1.6 micron, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.