missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt rod, 12.5mm (0.492in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 7440-48-4 |
Molecular Formula | Co |
MDL Number | MFCD00010935 |
Quantity | 25 mm |
Synonym | kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride |
InChI Key | GUTLYIVDDKVIGB-UHFFFAOYSA-N |
SMILES | [Co] |
IUPAC Name | cobalt |
Molecular Weight (g/mol) | 58.93 |
PubChem CID | 104730 |
Show More |
Safety and Handling
Product Identifier
- Cobalt rod
- Danger
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H413-May cause long lasting harmful effects to aquatic life.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction