missing translation for 'onlineSavingsMsg'
Learn More

Cobalt silicide, 99% (metals basis)

Product Code. 11352208
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11352208

Brand: Thermo Scientific Alfa Aesar 013115.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12017-12-8 | CoSi2 | 115.10 g/mol

Cobalt silicide is widely used in chemical research. It is also used for the production and characterization of very-large-scale integration (VLSI) applications in semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12017-12-8
Molecular Formula CoSi2
Molecular Weight (g/mol) 115.10
MDL Number MFCD00016021
InChI Key QKWVWIGBHQUEHX-UHFFFAOYSA-N

Specifications

Melting Point 1277°C
Color Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 115.11
Odor Odorless
Density 5.3
Chemical Name or Material Cobalt silicide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.