Learn More
Cobalt slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 25 g |
Percent Purity | 99.95% |
Odor | Odorless |
Weight | ≈0.22g/slug |
Form | Slug |
Assay | (metals basis) |
Chemical Name or Material | Cobalt slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M30L014 | Certificate of Analysis | 03/02/2025 | 042355.14,042355.06,042355.22 |
M06L003 | Certificate of Analysis | 23/01/2025 | 042355.14,042355.06,042355.22 |
T08K061 | Certificate of Analysis | 20/12/2024 | 042355.14,042355.06,042355.22 |
T08K062 | Certificate of Analysis | 06/09/2024 | 042355.14,042355.06,042355.22 |
T11J012 | Certificate of Analysis | 26/07/2023 | 042355.14,042355.06,042355.22 |
Safety and Handling


- Cobalt slug
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H360F-May damage fertility.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Cobalt
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.