missing translation for 'onlineSavingsMsg'
Learn More

Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni <0.15%, Thermo Scientific Chemicals

£105.53 - £442.20

Chemical Identifiers

CAS 12017-01-5
MDL Number MFCD00016022
Synonym Cobalt titanate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11328818
View Documents
Thermo Scientific Alfa Aesar
041670.18
50 g
£105.53
50g
Please to purchase this item. Need a web account? Register with us today!
11318818
View Documents
Thermo Scientific Alfa Aesar
041670.30
250 g
£442.20
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt titanium oxide shows magnetic properties at room temperature and used in electronics and as spintronic devices. It is used to improve the gas sensing ability of hydrogen sulfide by hydrothermal method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12017-01-5
MFCD00016022
Cobalt titanate

Specifications

12017-01-5
MFCD00016022
Insoluble in water.
99.8%
Cobalt titanium oxide
Powder
Cobalt titanate
154.83
(metals basis excluding Ni), Ni <0.15%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335-H350

missing translation for 'einecsNumber' : 234-615-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only