Learn More
Cobaltocene, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 1277-43-6 |
---|---|
Molecular Formula | C10H10Co |
Molecular Weight (g/mol) | 189.12 |
MDL Number | MFCD00013749 |
InChI Key | SNBRJOIYNQIWSZ-UHFFFAOYSA-N |
Synonym | Bis(cyclopentadienyl)cobalt |
PubChem CID | 24942376 |
IUPAC Name | cobalt(2+);cyclopenta-1,3-diene |
SMILES | C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1277-43-6 | |
189.12 | |
SNBRJOIYNQIWSZ-UHFFFAOYSA-N | |
24942376 | |
C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2] |
C10H10Co | |
MFCD00013749 | |
Bis(cyclopentadienyl)cobalt | |
cobalt(2+);cyclopenta-1,3-diene |
Specifications
1277-43-6 | |
Black | |
Authentic | |
C10H10Co | |
Bis(cyclopentadienyl)cobalt | |
SNBRJOIYNQIWSZ-UHFFFAOYSA-N | |
cobalt(2+);cyclopenta-1,3-diene | |
24942376 | |
98% | |
Cobaltocene |
173°C | |
97.5% min. | |
98% | |
MFCD00013749 | |
Solubility in water: insoluble.,soluble in hydrocarbons | |
C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2] | |
189.12 | |
189.12 | |
Crystals |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 215-061-
- Cobaltocene
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only