missing translation for 'onlineSavingsMsg'
Learn More

COD Standard Solution, 1000mg/L, Reagecon™
BIOPHARMA

Product Code. 12800317 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Verify that correct COD analysis has taken place. COD Standard Solution, 1000mg/L, Reagecon™ is a control and calibration standard that yields a precise COD measurement of 1000ppm. Achieving the anticipated result for the standard improves confidence in the COD readings for a sample.

Chemical oxygen demand (COD) is an important environmental parameter that measures the amount of oxygen that can be consumed by reactions in a measured solution. A COD test determines the amount of oxidizable pollutants in a water sample and helps determine the effect a particular effluent will have on a receiving body.

Specifications

Description COD control and calibration standard
Concentration 1000mg/L
Packaging HDPE Bottle
Quantity 500 mL
Traceability to NIST Yes
Odor Characteristic
Vapor Pressure 23 hPa
Density 1.00015g/cm3
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.