Learn More
Code Corporation™ Code Reader™ 5200 Barcode Scanner with Age Verification
Seamlessly connect your operations with the CR5200. The 5200 Barcode Scanner reads barcodes in any orientation, offers hands-free operation and can verify age by scanning ID cards.
Brand:  Code Corporation™ 176696
Product Code. 17983862
  • £577.00 / Each
Explore more special offers
Description
Description
  • Get high-speed, omnidirectional reading of 1D and 2D barcode symbologies
  • Features age verification when scanning ID cards
  • Quickly read barcodes on mobile device screens
  • Compact and adjustable for any reading direction
  • Provides user feedback with LED and audible tone
  • Features extended read ranges
Specifications
Specifications
Cable
USB
Barcode Scanner
139.7 mm
76.2 mm
88.9 mm
318 g
51° Horizontal by 39.4° Vertical
1 MB NOR Flash, 16 MB RAM
1280 x 960
BC412, Codabar, Code 11, Code 128, Code 32, Code 39, Code 93, GS1 DataBar, Hong Kong 2 of 5, IATA 2 of 5, Interleaved 2 of 5, MSI Plessey, Matrix 2 of 5, NEC 2 of 5, Plessey, Straight 2 of 5, Telepen, Trioptic, UPC/EAN/JAN, Pharmacode
5.5 in.
3 in.
3.5 in.
11.2 oz
CR5200, USB Cable