missing translation for 'onlineSavingsMsg'
Learn More

Melford Coelenterazine-H

Product Code. p-200087251
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 30018190

Brand: Melford C615000.001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly sensitive and specific luniophore used for measuring changes in levels of superoxide and Ca2+ in cells transfected with apoaequorin cDNA.

Exhibits 16-fold greater luminescence intensity as compared to native Coelenterazine.
  • CAS Number: 50909-86-9
  • Molecular Formula: C26H21N3O2
  • Molecular Weight: 407.5

Specifications

CAS 50909-86-9
Molecular Formula C26H21N3O2
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.