Learn More
Melford Coelenterazine Native
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion.
£133.00 - £401.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018202
|
Melford
C70900-0.001 |
1 mg |
£133.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018203
|
Melford
C70900-0.01 |
10 mg |
£401.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion.
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion. Used for detecting changes in intracellular Ca2+ in cells that have been transfected with apoaequorin cDNA.Specifications
55779-48-1 | |
C26H21N3O3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Melford Coelenterazine Native