missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Colcemid, 98+%

Product Code. 15456059
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-30-5
Molecular Formula C21H25NO5
Synonym Demecolcine; N-Methyl-N-desacetylcolchicine

Specifications

Quantity 50 mg
Description Pale yellow
Content And Storage 2°C to 8°C
For Use With (Application) Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
Formula Weight 371.43
Sensitivity Air and Light Sensitive
Melting Point 186°C
Solubility Soluble in 10mg/ml chloroform. Also soluble in ethanol
Form Powder
InChI Key NNJPGOLRFBJNIW-UHFFFAOYNA-N
SMILES CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12
IUPAC Name 3,4,5,14-tetramethoxy-10-(methylamino)tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-13-one
Molecular Weight (g/mol) 371.43
Assay 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'