Learn More
Thermo Scientific Chemicals Colchicine, 97%
Colchicine, 7alphah-colchicine, CAS # 64-86-8, is a laboratory grade formulation of the anti-inflammatory plant alkaloid drug.
£40.00 - £826.11
Chemical Identifiers
CAS | 64-86-8 |
---|---|
Molecular Formula | C22H25NO6 |
Molecular Weight (g/mol) | 399.44 |
MDL Number | MFCD00078484,MFCD00078484 |
InChI Key | IAKHMKGGTNLKSZ-INIZCTEOSA-N |
Synonym | colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine |
PubChem CID | 45038708 |
IUPAC Name | 2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide |
SMILES | COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10185230
|
Thermo Scientific Chemicals
227125000 |
500 mg | Glass bottle |
£40.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388310
|
Thermo Scientific Chemicals
|
1 g | Glass bottle |
£64.82
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174953
|
Thermo Scientific Chemicals
227120050 |
5 g | Glass bottle |
£208.04
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13428488
|
Thermo Scientific Chemicals
227120250 |
25 g | Glass bottle |
£826.11
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Colchicine (7alphah-colchicine) is an alkaloid found in Colchicum autumnale that has been shown to have anti-gout and anti-inflammatory properties
- Colchicine binds tubulin and prevents the migration of immune cells, reducing inflammation
Applications
- Colchicine is an anti-mitotic; may be used in the study of cell proliferation and migration
- Intended for in vivo and in vitro research applications
- For research use only, not intended to treat disease
Chemical Identifiers
64-86-8 | |
399.44 | |
IAKHMKGGTNLKSZ-INIZCTEOSA-N | |
45038708 | |
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1 |
C22H25NO6 | |
MFCD00078484,MFCD00078484 | |
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine | |
2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide |
Specifications
64-86-8 | |
Yellow | |
96% min. on anhydrous and solvent-free substance (HPLC) | |
MFCD00078484,MFCD00078484 | |
15,2455 | |
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine | |
IAKHMKGGTNLKSZ-INIZCTEOSA-N | |
2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide | |
45038708 | |
97% | |
Colchicine |
150.0°C to 160.0°C | |
Authentic | |
C22H25NO6 | |
14,IV,946 | |
− 250.00 | |
Solubility in water: 45g/l (20°C). Other solubilities: soluble in alcohol and chloroform,practically insoluble in petroleum ether,1g/220mL ether,1g/100mL benzene | |
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1 | |
399.44 | |
399.44 | |
Powder |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
May cause genetic defects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-598-5
- Colchicine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Germ cell mutagenicity Category 1B
Hazard Statement
- H340-May cause genetic defects.
- H300-Fatal if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only