missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs
Minimize prep time and maximize production for media fill trials with this ready-to-use soya broth in BioProcess Containers.
£367.00 - £1585.00
Specifications
Color | Clear, straw |
---|---|
Content And Storage | 2 to 25°C |
Description | Cold Filterable Tryptone Soya Broth in BPCs |
Format | Bioprocess Container |
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Includes | Price | Quantity & Availability | ||||
12980149
|
Thermo Scientific™
BP1065E |
20 L | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector |
£1585.00
20 L |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
12960149
|
Thermo Scientific™
BP1065A |
1 L | Includes the following connectors: LUER lock female +ö and SmartSite, needle-free valve additional sampling port |
£367.00
1 L |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
12970149
|
Thermo Scientific™
BP1065C |
10 L | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector |
£1010.00
10 L |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
Description
Minimize prep time and maximize production for media fill trials with ready-to-use Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs. Simply connect the BioProcess Containers into your existing workflow to maximize process simulations and minimize secondary contamination.
Effective- Achieve effective process simulations, with minimized risk of secondary contamination
- Just plug and play
- Minimize preparation time
- Satisfy regulatory compliance
- Save time, reduce labour costs
Easy-to-Use
- Innovative bag design and connector options for easy process integration
- Standard, ready-to-use, off-the-shelf product range
- No mixing, no sterilization, long shelf life at ambient storage
- No Mycoplasma contamination
- Qualified microbial recovery performance
- Comprehensive validation package for media, bag/components and filling process
- No filter blockages
- No mixing required
- No sterilization
- No WFI (water for injection) required
- No vessel cleaning
Pharmacopoeial-compliant Tryptone Soya Broth and Animal Derived Component Free (ADCF) vegetable peptone-based formulations are available.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
SmartSite™ is a trademark or registered trademark of the Cardinal Healthcare Inc.
ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.
Specifications
Clear, straw | |
Cold Filterable Tryptone Soya Broth in BPCs | |
Microbiology Media |
2 to 25°C | |
Bioprocess Container |