Learn More
Thermo Scientific™ Cold Filterable Vegetable Peptone Broth in BPCs
Minimize prep time and maximize production for media fill trials with this ready-to-use peptone broth, an alternative to TSB, in BioProcess Containers.
Brand: Thermo Scientific™ BP0104C
880.59 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 10.00000kg
Description
Minimize prep time and maximize production for media fill trials with ready-to-use Thermo Scientific™ Cold Filterable Vegetable Peptone Broth in BPCs, a vegetable based alternative to TSB in BioProcess Containers. Simply connect the BPCs into your existing workflow to maximize process simulations and minimize secondary contamination.
Effective- Achieve effective process simulations, with minimized risk of secondary contamination
- Just plug and play
- Minimize preparation time
- Satisfy regulatory compliance
- Save time, reduce labour costs
- Innovative bag design and connector options for easy process integration
- Standard, ready-to-use, off-the-shelf product range
- No mixing, no sterilization, long shelf life at ambient storage
- No Mycoplasma contamination
- Qualified microbial recovery performance
- Comprehensive validation package for media, bag/components and filling process
- No filter blockages
- No mixing required
- No sterilization
- No WFI (water for injection) required
- No vessel cleaning
Specifications
Clear, straw | |
Cold Filterable Vegetable Peptone Broth in BPCs | |
Microbiology Media | |
1/Pk. |
2°C to 8°C | |
Bioprocess Container | |
Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector | |
10 L |