Learn More
VACUUBRAND™ Cold Trap
Use this Cold Trap with very volatile solvents and reduce the back streaming of oil molecules into the vacuum system. Vacubrand™ Cold Traps provide important protection for your pump by separating condensates and aggressive media.
Brand: VACUUBRAND™ 20667051
Code : A0
Additional Details : Weight : 0.80000kg
Description
Cold trap SKF- Sturdy, easy to clean
- Easy to disassemble
- Two-wall design with good conductance
- Long operating time per coolant filling
- Easy condensate drainage without disassembling
Cold trap GKF
- Highly reflective insulation jacket for extended coolant life
- Vertical window: Direct observation of condensate and coolant levels
- PTFE stopcock: Drain condensate without disassembly
- Sheet metal shield for protection against external damage and implosion
Inline oil filter HF
- Extended oil lifetime
- Reduced service demands
- Easy space-saving mounting
- Simple filter change, service indicator
Specifications
Stainless Steel, FKM, NBR | |
Vacuubrand pumps | |
SKF H 25 Cold Trap |