missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ BioPharm Plus™ L/S™ Precision Pump Tubing, Platinum-Cured Silicone
Ensure optimal performance from your Masterflex™ pump with this platinum-cured silicone tubing. Masterflex™ BioPharm Plus™ L/S™ Precision Pump Tubing is ideal for pharmaceutical, biotechnology, peristaltic pumping and food applications.
Brand: Masterflex™ 96440-25
This item is not returnable.
View return policy
Description
- Ideal for pharmaceutical and biotechnological applications
- Low extractable, excellent heat resistance and low compression set
- Custom extruded to fit Masterflex™ pumps
- Provides longer life than other platinum and peroxide silicone tubing formulations
- Manufactured according to Good Manufacturing Practices (GMP)
- Longer tubing life means less tubing changes and maintenance requirements
- Complies with USP Class VI, EP, FDA, 3A, REACH, RoHS, ADCF
- Excellent biocompatibility and low absorption
- Translucent, odorless and nontoxic
- Weather, ozone, corona and radiation resistant
- Can be sterilized by autoclave, gamma radiation or ethylene oxide
Specifications
USP Class VI, FDA 21 CFR 177.2600, Exceeds 3A sanitary standards, European Pharmacopoeia (EP) | |
Autoclavable | |
4.8 mm | |
25 ft. | |
pharmaceutical, biotechnology, peristaltic pumping, and food applications | |
25 | |
3/16 in. | |
8.8 m | |
Do not use with concentrated solvents, oils, acids. Relatively high gas permeability. | |
35 psi | |
232°C |
Pump Tubing | |
3/16 in. | |
Thermal-set Rubber, Siloxane Polymers, Amorphous Silica | |
760 cm | |
-59°C | |
21 to 105mL/min. | |
Sterilize by ETO, autoclave, or gamma radiation up to 2.5 Mrad. To autoclave: coil loosely in nonlinting cloth or paper; autoclave at 121┬░C (250┬░F), 1 bar (15 psi) for 30 minutes. | |
CO2: 25,147; H2: ----; O2: 4715; N2: 2284 | |
Translucent | |
660 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction