missing translation for 'onlineSavingsMsg'
Learn More
Masterflex™ Nylon Male Luer Fitting Product Code.: 11868621
Only undefined left
Add to basket

Masterflex™ Nylon Male Luer Fitting

Product Code. 11868621
Male Luer w/ Lock Ring x 3/16 in. Hose Barb, Pack of 25
missing translation for 'orderingAttributeHoverText'
For Use With:
Male LL to 1/16 in. I.D.
Male Luer to 1/4 to 28 in. Thread
Male Luer w/ Lock Ring x 1/16 in. Hose Barb
Male Luer w/ Lock Ring x 1/4 in. Hose Barb
Male Luer w/ Lock Ring x 1/8 in. Hose Barb
Male Luer w/ Lock Ring x 3/16 in. Hose Barb
Male luer x 1/4 in. hose barb
missing translation for 'unitSize'
Pack of 25
This item is not returnable. View return policy

Product Code. 11868621

Brand: Masterflex™ 4550508

Only undefined left
Add to basket
Only undefined left
Add to basket
This item is not returnable. View return policy

Ensure process integrity with full material traceability. Masterflex™ Nylon Male Luer Fittings are ideal for a variety of critical applications, including syringes or small tubing. These fittings are inspected to be free from contaminants, and are fully traceable.

  • Meet tight dimensional tolerances for increased reliability and process purity
  • Choose from a range of sizes, 1⁄16" to 1⁄4", with a variety of connection options
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • In compliance with ISO 594-1 standards
  • Good chemical resistance
  • Max temperature 455°F (235°C)
  • Sterilize by autoclave, ethylene oxide, or gamma irradiation

Specifications

Certifications/Compliance ISO 9001
Material Nylon
For Use With Male Luer w/ Lock Ring x 3/16 in. Hose Barb
Packaging Quantity 25 Pack
Includes Wide-bore hose barb for enhanced fluid path flow
Product Type Luer Fitting
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Masterflex™ Nylon Male Luer Fitting >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.