Learn More
3M™ Petrifilm™ Coliform Count Plates
Sample-ready to identify coliform levels in raw materials, process/finished food products and plant environment/equipment
Brand:  3M™ Petrifilm™ 7100126806
Encompass
Product Code. 17203784
  • £1185.15 / Pack of 1000
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Coliform results in 24 hours.
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
  • Interpretation can be done with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
  • This plate has been awarded Official Methods of Analysis (OMA) #986.33 for Milk, OMA #989.10 for dairy products and OMA #991.14 for Food NF VALIDATION certificates: 3M 01/02-09/89 A for all human food products (except raw shellfish), Pet Food and Environmental samples, 3M 01/02-09/89 B for all human food products (except raw shellfish), and 3M 01/02-09/89 C for all human food products by AFNOR Certification
  • Food processors and Contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Environmental Test
1000