missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Colistin sulfate salt Product Code.: 15565146

Thermo Scientific Chemicals Colistin sulfate salt

Product Code. 15565146
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15565146

Brand: Thermo Scientific Chemicals 455390010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colistin Sulfate salt (2:5), CAS # 1264-72-8, (synonym: Polymyxin E Complex), is a complex mixture of antibiotics, with Colistin A and B being the primary components. It has anti-bacterial activity against Gram-positive and Gram-negative bacteria and may also be used as a neuromuscular blocker.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Colistin Sulfate salt is a stable, cationic, water-soluble, white powder composed of over 30 components
• Originally isolated from B. polymyxa, it inhibits the growth of the Gram-negative (e.g. E. coli, P. aeruginosa, P. fluorescens, and S. enterica), and Gram-positive bacteria (e.g. L. lactis, P. polymyxa, P. acidilactici, and S. aureus) by permeabilizing the bacterial cell membrane

Applications
• Colistin Sulfate has potent anti-endotoxin activity rendered through its ability to bind lippopolysaccharides that make up bacterial endotoxins
• This compound is often used in the management of multi-drug resistant Gram-negative infections

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264-72-8

Specifications

Type Colistin Sulfate salt
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in methanol, insoluble in acetone, diethyl ether
Percent Purity ≥77% Sum of Polymyxins (HPLC)
Chemical Name or Material Colistin sulfate salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0445144 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number455390010,455392500
Lot NumberA0451458 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number455390010,455392500
Lot NumberA0434721 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number455390010,455392500
Lot NumberA0445144 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number455390010,455392500
Lot NumberA0376967 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number455390010,455392500
compliance-icons
Product Identifier
  • Colistin sulfate salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Colistin sulfate salt >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.