missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Collagen I, rat tail

Product Code. 11519816
Click to view available options
Quantity:
25 mL
Unit Size:
20mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11519816

Brand: Gibco™ A1048301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Collagen I Rat Protein, Tail

Collagen I, Rat Tail is the most common fibrillar collagen (90%) found in skin, bone, tendons, and other connective tissues. It is supplied at ~4 mg/mL in 20 mM acetic acid, providing the flexibility to dilute to lower concentrations. (See Certificate of Analysis for exact concentration.) The detailed gelling procedure forms a firm, clear gel for 3D applications.

Collagen is a fibrous protein that consists of three α-chains that can combine to form a rope-like triple helix, providing tensile strength to the extracellular matrix (ECM) where it plays a key role in cell growth, differentiation, attachment, and cell migration. The α-chains contain GXY repeats. Glycine (G) is a small amino acid that fits well in the triple helix. X and Y are typically proline and hydroxyproline, which is critical for collagen stability. Type I is the most common fibrillar collagen (90%) and is mostly found in skin, bone, tendons, and other connective tissues.

Rat tail collagen I can be prepared as a clear gel providing a 3D matrix or surface coated on tissue culture plates as a substrate for culturing primary cells such as keratinocytes and hepatocytes.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4 mg/mL
For Use With (Application) Cell Culture
Quantity 25 mL
Product Type Collagen
Species Rat
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Endotoxin Level ≤10 EU/mL, ≤ 10 EU/mL
Shipping Condition Wet Ice
Protein Form Native
Form Liquid
Protein Subtype Collagen
Research Category Extracellular Matrix & Cell Adhesion
Product Line Gibco
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.