Learn More
SymLine Collector, V2.0, 3/4-ways
This purging manifold allows you to install up to four SafetyCaps on your purging bottle.
Brand: SymLine 306509
202.70 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.25300kg
Description
- Safe and easy handling - Capillaries are reliably fixed in place
- Solvent filters have sufficient space due to the wide GL45 opening
- Unused connections can be easily closed with the supplied blank cap
The system for Solvent Safety remains permanently closed, the safety chain remains intact during the rinsing process.
Specifications
Universal 3/4-ways incl. blind cap for HPLC connection, 1 x GL45 (f) to 4 x GL45 (m) | |
Safety Waste Cap | |
SymLine System | |
Collector, V2.0, universal 3/4-ways incl. blind cap for HPLC connection, 1 x GL45 (f) to 4 x GL45 (m), PE-HD electrostatic conductive |
Waste Disposal System | |
HDPE-EC | |
GL45 |