missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience Scan 500 Automatic Colony Counter
Scan 500 is an automatic colony counter that ensures fast and efficient counting of colonies, including on chromogenic agar. It provides excellent reproducibility.
Brand: Interscience 436000
This item is not returnable.
View return policy
Description
Automatic colony counter & inhibition zone reader, delivered with software.- Counting on Petri dishes (55 - 90 mm)
- Spiral™ plating
- Color detection: 7 colors on the same dish
- Counting on chromogenic medium
- Inhibition zone measurement
Specifications
Automatic Colony Counter | |
CMOS color camera, M12 lens, Zoom x 28, Resolution: 1 megapixel | |
USB | |
28.7 x 27.1 x 29 cm | |
11.42 in. | |
8.38 kg | |
100/240 V | |
21 CFR Part 11, ISO 7218 and AOAC 977.27 | |
Counting on round Petri Dishes of dia. 55 - 90 mm, PetriFilmTM, MC-Media PadsTM, Compact DryTM, easyPlateTM, filtration membranes, Compatible with dataLinkTM or dataLinkTMpro,traceability system |
USB | |
Stainless Steel | |
Counting time: up to 1000 colonies per second, Number of antibiotic paper disks: up to 7 antibiotics on a dia. 90 mm Petri dish, Reading time: 7 inhibition zone reading between 1 to 3 seconds | |
10.67 in. | |
11.30 in. | |
50-60 Hz | |
Max power: 20 W | |
The Scan 500 colony counter is used in the food, environmental, pharmaceutical, cosmetics, veterinary and public institutes research. | |
1 Scan™ software, 1 15 V power cord, 1 USB cable, 3 validation plates, 1 user manual |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Interscience Scan 500 Automatic Colony Counter > For use with: Petri dishes
Spot an opportunity for improvement?Share a Content Correction