missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP ColonyDoc-It Colony Counter
Capture and count a wide array of white light and fluorescently-marked colonies. UVP Ultra Violet Product™ ColonyDoc-It Colony Counter uses software that enables fast, simple colony counting.
Brand: UVP 97-0539-02
This item is not returnable.
View return policy
Description
- DigiCam 130 digital color camera with 17.9 megapixel resolution
- Select from several light sources for illuminating a wide range of stained media
- Slide the filter selector to one of two positions
- A wide range of emission filters are available
- The plate alcove accommodates pour, spread and spiral plates and filters with sizes from 33mm to 150mm
- Capture colony sizes as small as 0.08mm
- Generate fast automatic and accurate colony counting with detailed statistics
- Define parameters including eight color differentiation, split or merge, filter by group or size
- Attach the door (not shown) to create a darkroom environment when imaging colonies with GFP fluorescence
- Visualize a wide array of bacterial, yeast and mold colonies with samples found in air, water, food and cosmetics.
- Darkfield
- Epi and transillumination white light
- Epi blue light
- Integrated 17.9 megapixel digital color camera captures images down to 0.08mm.
- Provides camera control, image capture and analysis.
- Generates statistics and displays the most critical information about the colony area, perimeter, average diameter and circularity. The data is easily exported into Excel.
Specifications
Colony Counter | |
343 x 318 x 445mm | |
230V |
Lighting: Epi White, Transillumination White, Epi Blue, Darkfield | |
20lbs | |
17.9MP Camera |