Learn More
Shamrock™ Color Coded Multi-Purpose Labeling Tape
Offered in a number of colors and sizes to fit your needs. Shamrock™ Color Coded Multi-Purpose Labeling Tape offers multiple opportunities for color coding in the laboratory - test, physician or department.
£6.17 - £158.00
Specifications
Adhesive Type | Removable |
---|---|
Product Type | Color-coded Multi-purpose Labeling Tape |
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
11731596
|
Shamrock™
ST-11-6 |
Blue | 12.7 m x 38 mm |
£14.69
12m |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12320190
|
Shamrock™
ST-10-RNBW |
Assorted | 12.7 m x 25 mm |
£158.00
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12310190
|
Shamrock™
ST-12-RNBW |
Assorted | 12.7 m x 12.7 mm |
£122.00
Pack of 24 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11701616
|
Shamrock™
ST-20-6 |
Blue | 12.7 m x 50 mm |
£16.90
12m |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11700085
|
Shamrock™
ST-10-23 |
Brown | 12.7 m x 25 mm |
£10.35
12m |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11867193
|
Shamrock™
ST-12-17 |
Chartreuse | 12.7 m x 12.7 mm |
£6.17
12m |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11879070
|
Shamrock™
ST-34-10 |
Copper | 55 m x 19 mm |
£9.00
12m |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11827193
|
Shamrock™
ST-12-10 |
Copper | 12.7 m x 12.7 mm |
£6.85
12m |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11740075
|
Shamrock™
ST-12-23 |
Dark Brown | 12.7 m x 12.7 mm |
£6.20
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11770075
|
Shamrock™
ST-34-23 |
Dark Brown | 55 m x 19 mm |
£8.12
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11701596
|
Shamrock™
ST-11-25 |
Green | 12.7 m x 38 mm |
£14.69
12m |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11790075
|
Shamrock™
ST-10-14 |
Gray | 12.7 m x 25 mm |
£10.35
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11771606
|
Shamrock™
ST-20-3 |
Green | 12.7 m x 50 mm |
£18.60
12m |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11721596
|
Shamrock™
ST-11-5 |
Orange | 12.7 m x 38 mm |
£14.69
12m |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11791606
|
Shamrock™
ST-20-5 |
Orange | 12.7 m x 50 mm |
£16.90
12m |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11887203
|
Shamrock™
ST-34-13 |
Lavender | 12.7 m x 19 mm |
£10.40
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11897203
|
Shamrock™
ST-34-15 |
Lime Green | 12.7 m x 19 mm |
£8.95
12m |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11711616
|
Shamrock™
ST-20-7 |
Pink | 12.7 m x 50 mm |
£16.90
12m |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11711596
|
Shamrock™
ST-11-4 |
Red | 12.7 m x 38 mm |
£14.69
12m |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11877203
|
Shamrock™
ST-34-11 |
Rose | 55 m x 19 mm |
£8.12
12m |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11721606
|
Shamrock™
ST-11-12 |
Violet | 12.7 m x 38 mm |
£14.69
12m |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11869070
|
Shamrock™
ST-34-8 |
Tan | 55 m x 19 mm |
£8.95
12m |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11791586
|
Shamrock™
ST-11-2 |
Yellow | 12.7 m x 38 mm |
£14.70
12m |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11761606
|
Shamrock™
ST-20-2 |
Yellow | 12.7 m x 50 mm |
£16.90
12m |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11781586
|
Shamrock™
ST-11-1 |
White | 12.7 m x 38 mm |
£16.20
12m |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Resistant to water, oil and acid
- Temperature range of -20 to +121°C
- For autoclave, freezer, refrigerator and oven
- Universal marking with ballpoint pen, paper pencil and felt marker
Specifications
Removable | |
Color-coded Multi-purpose Labeling Tape |