missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Blood Agar with 8mg/L Aztreonam

Product Code. 12937148
Click to view available options
Description:
Columbia Blood Agar with 8mg/L Aztreonam
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12937148

Brand: Thermo Scientific™ PB0835A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate and isolate a wide variety of fastidious microorganisms with this multi-purpose, ready-to-use medium.

Cultivate and isolate a wide variety of fastidious microorganisms with Thermo Scientific™ Oxoid™ Columbia Blood Agar with 8mg/L Aztreonam. Columbia Agar is an ideal base medium for the addition of blood, especially in selective formulations when supplemented with antimicrobial agents. The addition of defibrinated horse blood and Aztreonam, in this case, facilitates the selective cultivation of Gram-positive fastidious microorganisms.

Traditionally, blood agar bases have been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies and of the second in clearly defined zones of hemolysis and good colonial differentiation. Columbia Blood Agar with 8mg/L Aztreonam combines the virtues of both of these types of media to give an improved all-round performance, showing versatility and superior performance in several applications.1

  • Produces large colonies of fastidious microorganisms with clearly defined hemolytic reactions and colonial morphology.
  • Selects for Gram-positive bacteria with the addition of Aztreonam.
  • Ready-to-use convenience of prepared media

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage 2°C to 10°C
Description Columbia Blood Agar with 8mg/L Aztreonam
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.