Learn More
Thermo Scientific™ Columbia Horse Blood CNA Agar
Identify Gram-positive cocci from clinical specimens and food samples with this selective medium for staphylococci and streptococci.
Brand: Thermo Scientific™ PB0308A
15.36 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.30000kg
Description
Recognize Gram-positive cocci easier using Thermo Scientific™ Columbia Horse Blood CNA Agar, a selective medium for the isolation of staphylococci and streptococci from both clinical specimens and food samples.
Columbia CNA Agar with Horse Blood contains antibiotics to inhibit S. albus and Micrococcus species as well as Gram-positive and Gram-negative rods. It suppresses growth of Proteus, Klebsiella and Pseudomonas species while permitting unrestricted growth of S. aureus, haemolytic streptococci and enterococci. This medium enables important Gram-positive cocci to be recognised more readily and isolated easily from the mixed bacterial populations contained in many clinical specimens and foods.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Spécification
Red, opaque | |
Columbia Horse Blood CNA Agar | |
United Kingdom | |
19.5 g ± 1.0 g | |
10/Pk. |
2°C to 10°C | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |