Learn More
Cytiva Column Holder Rod
Product Code. 12415630 Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
HiTrap and Tricorn Columns
Unit Size:
Each
Product Code. 12415630
Brand: Cytiva 28956270
Only null left
Add to basket
Add to basket
Holds up to five columns
Holder for up to five HiTrap columns for ÅKTA avant and ÅKTA pure.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) HiTrap and Tricorn Columns
Type Column Holder Rod
Quantity 1 Each