Learn More
Macherey-Nagel™ Columns for NucleoSpin™ RNA Plus
Additional columns for NucleoSpin™ RNA Plus kits for quick isolation of RNA from cells and tissues. gDNA Removal Columns enable lysate clearing and removal of genomic DNA in one step on the same column; RNA Binding Columns efficiently isolate purified RNA.
Brand:  Macherey-Nagel™ 740984.50S
184.59 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 15820978
  • £256.00 / Pack of 50
Explore more special offers
Description
Description
NucleoSpin™ RNA Plus
  • Efficient and quick RNA isolation from cells and tissues
  • Two-column system includes gDNA removal columns and RNA binding columns
  • NucleoSpin™ gDNA Removal Columns – no time consuming rDNase digestion
  • Convenient handling – lysate clearing and gDNA removal on one column in one step
  • New efficient lysis buffer – no b-mercaptoethanol or TCEP necessary
  • Variety of sample materials – cultured cells, animal tissue, plant tissue, microorganisms
  • Downstream applications include real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Specifications
Specifications
Blue ring
Mini Spin Column
30 - 120 μL
Binding capacity: 200 μg
NucleoSpin™ RNA Plus Kits
RNA Plus Binding Column
< 30 mg tissue; < 30 mg tissue
Room temperature (18-25 °C)
Silica Membrane
Cultured Bacterial or Yeast Cells, Human or Animal Tissue
RNA (with typical RIN > 9)
Isolation of RNA
50
20 min/6 preps
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • NucleoSpin RNA Plus
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : ethyleneglycol diacetate / guanidinium thiocyanate /guanidine hydrochloride, ethanol