Learn More
Lovibond™ Combi pack AMMONIA #1 / #2
Tablet reagent test set
Brand: Lovibond™ 517611BT
Code : NEW
Additional Details : Weight : 0.01400kg Transport : UN number : 2680 Chem class : 8 Pack group : II
Description
Tablet reagent test set
Combi pack AMMONIA #1 / #2 100 tests measuring range 0.02 - 1.00 mg/L Indophenol blue method for comparators and photometers.
Specifications
100 Tests | |
AMMONIA #1 to #2, 0.02 to 1.00 mg/L | |
Reagent |
Safety and Handling
- combi-pack ammonia No. 1 + No. 2
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : lithium hydroxide monohydrate, sodium dichloroisocyanurate,dihydrate, salicylicacid, sodium nitrop