Learn More
Lovibond™ Combi pack COPPER #1 / #2
Tablet reagent test set
Brand:  Lovibond™ 517691BT
Product Code. 15331925
  • £114.00 / Pack of 100
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Tablet reagent test set
Combi pack COPPER #1 / #2 100 tests measuring range 0.05 - 5.00 mg/L Biquinoline method for photometers. 
Specifications
Specifications
100 Tests
COPPER #1 to #2 , 0.05 to 5.00 mg/L
Reagent
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • combi-pack copper No. 1 + No. 2
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
Supplemental information
  • MIXTURE LIST-Contains : calcium thioglycolate trihydrate, adipicacid