Learn More
Combination Semi-Volatile Surrogate Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CLPS-SURR
Code : A0
Additional Details : Weight : 0.00100kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
2,4,6-Tribromophenol | 118-79-6 | 2000μg/mL |
2-Fluorophenol | 367-12-4 | 2000μg/mL |
Phenol-d6 | 13127-88-3 | 2000μg/mL |
2-Fluorobiphenyl | 321-60-8 | 1000μg/mL |
Nitrobenzene-d5 | 4165-60-0 | 1000μg/mL |
p-Terphenyl-d14 | 1718-51-0 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Combination Semivolatile Surrogate Standard | |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For US EPA CLP Methods |
CLP SOW 2/88 | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
MeCl:Acetone |
Safety and Handling
- Combination Semivolatile Surrogate Standard
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dichloromethane, acetone, nitrobenzene-d5
Product intended for laboratory use only