missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™ MS20-2-0.75
Description
- Recommended for high resolution gels and publication quality data
- For tightly resolved bands
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
Black | |
Comb | |
1 Each | |
236 μL |
MultiSUB Maxi-Gel Electrophoresis Unit | |
2 Prep, 2 Marker | |
0.75 mm |